missing translation for 'onlineSavingsMsg'
Läs mer

Nonanal, Sigma-Aldrich™

Produktkod. 15879430
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mL
Förpackningsstorlek:
50ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15879430 50 mL 50ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15879430 Leverantör Sigma Aldrich Leverantörsnummer 8071660050
 Begränsad artikel
Beräknad leverans: 30-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 124-19-6
Densitet 0.83 g/cm3 at 20°C
Molekylformel CH3(CH2)7CHO
MDL-nummer MFCD00007030
Kvantitet 50 mL
Synonym Nonanal, Pelargonaldehyde, Nonanal
Molekylvikt (g/mol) 142.24
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.