Läs mer
Nonidet P 40 Substitute, TRC
Produktkod. 30466093
Change view
Kvantitet:
1 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466093 1 g
30467083 100 g
30469646 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30466093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN6495701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 9016-45-9
Molekylformel C15H24O.(C2H4O)n
Molekylvikt (g/mol) 308.5
Synonym α-(Nonylphenyl)-ω-hydroxypoly(oxy-1,2-ethanediyl), Nanophenoxypoly(ethyleneoxy)ethanol, (Nonylphenoxy)polyethylene oxide, A 730, A 730 (surfactant), Ace Clean AD, Adekanol NP 1000, Adekatol NP, Adekatol NP 1000, Adekatol NP 1100, Adekatol NP 638, Adekatol NP 650, Adekatol NP 660, Adekatol NP 675, Adekatol NP 683, Adekatol NP 686, Adekatol NP 690, Adekatol NP 700, Adekatol NP 710, Adekatol NP 720, Adekatol NP 760, Adekatol NP 900, Afilan CVH, Agral, Agral 600, Agral 90, Agral LN, Agral Plus, Agral R, Tensioactiv NF 10, Tensioactiv NF 6, Tenzilin 080, Tenzilin FN 65, Tergitol 9.5, Tergitol N 4, Tergitol N 8, Tergitol NO 10, Tergitol NP, Tergitol NP 10, Tergitol NP 101, Tergitol NP 12, Tergitol NP 13, Tergitol NP 14, Tergitol NP 15, Tergitol NP 27, Tergitol NP 30, Tergitol NP 33, Tergitol NP 35, Tergitol NP 4, Tergitol NP 40, Tergitol NP 6, Tergitol NP 7, Tergitol NP 70, Tergitol NP 8, Tergitol NP 9, Tergitol NPX, Tergitol TH, Tergitol TNP 10, Tergitol TP 9, Teric GN, Teric GN 5, Teric GN 8, Teric GN 9, Teric N, Teric N 100, Teric N 12, Teric N 13, Teric N 15, Teric N 2, Teric N 30, Teric N 300, Teric N 40, Teric N 450, Teric N 5, Teric N 8, Teric N 9, Texofor FN 6, Texofor FN 8, Triton N, Triton N 100, Triton N 101, Triton N 111, Triton N 128, Triton N 150, Triton N 302, Triton N 401, Triton N 42, Triton N 57, Triton N 60, Triton N 998, Trycol 6940, Trycol 6954, Trycol 6961, Trycol 6964, Trycol 6968, Trycol 6969, Trycol 6970, Trycol 6974, Trycol NP 30, Trycol NP 40, Ucefal DCN, Ultranex NP 100, Ultranex NP 1000, Ultranex NP 50, Ultranex NP 500, Ultranex NP 95, Unitop 100, Unitop 50, Value 3706, Varonic N 30-7, Varonic N 6, Veranol H 10, Veranol N 10, Vibnon 45, Wasc, Wellaid 711W, Weranol H 10, Witconate NP 120, Witconol NP 100, Witconol NP 120, Witconol NP 40, Witconol NP 60, Witconol NP 80, YF 6500, α-(Nonylphenyl)-ω-hydroxypoly(oxy-1,2-ethanediyl), α-(Nonylphenyl)-ω-hydroxypolyoxyethylene, ω-Hydroxy-α-(nonylphenyl)poly(oxy-1,2-ethanediyl)
LEDER OCCOC1=CC=C(CCCCCCCCC)C=C1
Specifikationer
Kemiskt namn eller material Nonidet P 40 Substitute
Kvantitet 1 g
InChI formel InChI=1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3