Läs mer
(-)-Nopol, TRC
Produktkod. 30480353
Change view
Kvantitet:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480353 5 g
30479313 100 g
30479319 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN6506305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35836-73-8
Molekylformel C11H18O
Molekylvikt (g/mol) 166.26
Synonym (1R,5S)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol, (1R)-(-)-Nopol, (1R)-(-)-Nopyl Alcohol, (1R)-Nopol
IUPAC-namn 2-[(1S,5R)-6,6-dimethyl-4-bicyclo[3.1.1]hept-3-enyl]ethanol
LEDER CC1(C)[C@H]2CC=C(CCO)[C@@H]1C2
Specifikationer
Kemiskt namn eller material (-)-Nopol
Kvantitet 5 g
InChI formel InChI=1S/C11H18O/c1-11(2)9-4-3-8(5-6-12)10(11)7-9/h3,9-10,12H,4-7H2,1-2H3/t9-,10-/m0/s1
Formel vikt 166.1358