Learn More
Nor Doxepin Hydrochloride, TRC
Product Code. 30424141
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424141 10 mg 10mg
30411622 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30424141 Supplier Toronto Research Chemicals Supplier No. TRCN67350010MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nordoxepin Hydrochloride
CAS 2887-91-4
Purity Grade Notes HPLC
Molecular Formula C18 H19 N O . HCl
Quantity 10 mg
Synonym 1-Propanamine','3-dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-','hydrochloride (1:1), 1-Propanamine','3-dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-','hydrochloride (9CI), Dibenz[b,e]oxepin-Δ11(6H),γ-propylamine','N-methyl-','hydrochloride (7CI,8CI), Dibenz[b,e]oxepin','1-propanamine deriv., Desmethyldoxepine hydrochloride, N-Desmethyldoxepin hydrochloride, Nordoxepin hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine hydrochloride, Nordoxepin hydrochloride (cis/trans), N-Desmethyldoxepin hydrochloride (cis/trans), Doxepin-desmethyl hydrochloride
InChI Formula InChI=1S/C18H19NO.ClH/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18;/h2-5,7-11,19H,6,12-13H2,1H3;1H
SMILES Cl.CNCCC=C1c2ccccc2COc3ccccc13
IUPAC Name 3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N-methylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 301.81
Formula Weight 301.123
Percent Purity >95