Läs mer
Nor NOHA monoacetate, Tocris Bioscience™
Arginase inhibitor
Brand: Tocris Bioscience 6370/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Nor NOHA monoacetate is a reversible, competitive arginase inhibitor (IC50 = 2 μM). Exhibits 10-fold selectivity for human type II arginase over type I. Enhances the effect of acetylcholine on isolated aortic and mesenteric arterial rings. Inhibits growth of lung carcinoma implants in mice.Specifikationer
| (2S)-2-Amino-4-[[(hydroxyamino)iminomethyl]amino]butanoic acid monoacetate | |
| 2250019-93-1 | |
| 10 mg |