Läs mer
Nor Propranolol Hydrochloride, TRC
Produktkod. 30423534
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30423534 5 mg
1 options
Produktkod. 30423534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN8256005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Nor Propranolol Hydrochloride
CAS 62618-09-1
Molekylformel C13 H15 N O2 . Cl H
Kvantitet 5 mg
Synonym 1-Amino-3-(1-naphthalenyloxy)-2-propanol Hydrochloride, 1-Amino-3-(α-naphthyloxy) propan-2-ol Hydrochloride, (+/-)-Desisopropylpropranolol Hydrochloride, (+/-)-Norpropranolol Hydrochloride, N-Deisopropylpropranolol Hydrochloride, N-Desisopropylpropranolol Hydrochloride
InChI formel InChI=1S/C13H15NO2.ClH/c14-8-11(15)9-16-13-7-3-5-10-4-1-2-6-12(10)13;/h1-7,11,15H,8-9,14H2;1H
LEDER Cl.NCC(O)COc1cccc2ccccc12
IUPAC-namn 1-amino-3-naphthalen-1-yloxypropan-2-ol;hydrochloride
Molekylvikt (g/mol) 253.7246
Formel vikt 253.087