Läs mer
Norantipyrine Glucuronide Sodium Salt, TRC
Produktkod. 30481222
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30481222 10 mg
1 options
Produktkod. 30481222 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN66110510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Norantipyrine Glucuronide Sodium Salt
CAS 663618-35-7
Molekylformel C16H17N2NaO7
Kvantitet 10 mg
Synonym 3-Methyl-1-phenyl-1H-pyrazol-5-yl beta-D-glucopyranosiduronic Acid Sodium Salt, MCI 186 Glucuronide Sodium Salt
InChI formel InChI=1S/C16H18N2O7.Na/c1-8-7-10(18(17-8)9-5-3-2-4-6-9)24-16-13(21)11(19)12(20)14(25-16)15(22)23;/h2-7,11-14,16,19-21H,1H3,(H,22,23);/q;+1/p-1/t11-,12-,13+,14-,16+;/m0./s1
LEDER [Na+].Cc1cc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)[O-])n(n1)c3ccccc3
IUPAC-namn sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(5-methyl-2-phenylpyrazol-3-yl)oxyoxane-2-carboxylate
Molekylvikt (g/mol) 372.31
Formel vikt 372.0933