Läs mer
Norantipyrine Sulfate Sodium Salt (>90%), TRC
Produktkod. 30480587
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30480587 25 mg
1 options
Produktkod. 30480587 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN66109525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Norantipyrine Sulfate Sodium Salt
CAS 663618-36-8
Molekylformel C10H9N2NaO4S
Kvantitet 25 mg
Synonym MCI 186 sulfate Sodium Salt, 3-Methyl-1-phenyl-1H-Pyrazol-5-ol Hydrogen Sulfate Sodium Salt (Ester)
InChI formel InChI=1S/C10H10N2O4S.Na/c1-8-7-10(16-17(13,14)15)12(11-8)9-5-3-2-4-6-9;/h2-7H,1H3,(H,13,14,15);/q;+1/p-1
LEDER [Na+].Cc1cc(OS(=O)(=O)[O-])n(n1)c2ccccc2
IUPAC-namn sodium;(5-methyl-2-phenylpyrazol-3-yl) sulfate
Molekylvikt (g/mol) 276.24
Formel vikt 276.0181