Läs mer
Noratropine Hydrochloride, TRC
Produktkod. 18070640
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18070640 100 mg
18032240 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18070640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN661080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75559-01-2
Molekylformel C16 H21 N O3 . H Cl
Molekylvikt (g/mol) 311.8
Synonym Noratropine hydrochloride solution, Noratropine HCl;
LEDER Cl.CC(=O)OCC[C@H](N)C(=O)O
Specifikationer
Kemiskt namn eller material Noratropine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C16H21NO3/c18-10-15(11-4-2-1-3-5-11)16(19)20-14-8-12-6-7-13(9-14)17-12/h1-5,12-15,17-18H,6-10H2/t12-,13+,14?,15-/m1/s1
Formel vikt 275.1521
Procent renhet >95%