Läs mer
Norclozapine.HCl, 1mg/ml in Isopropanol (as free base), Lipomed
Beskrivning
Norclozapine.HCl, 1mg/ml in Isopropanol (as free base)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Norclozapine Hydrochloride |
| CAS | 2741767-92-8 |
| Analyt- eller komponentnamn | Norclozapine Hydrochloride |
| Frakt skick | Ice Pk. (-20°C) |
| Molekylformel | C17 H17 Cl N4 . Cl H |
| Koncentration | 1 mg/mL |
| Synonym | 5 H-Dibenzo[b,e][1,4 ]diazepine, 8-chloro-11-(1-piperazinyl)-, hydrochloride (1:1) (ACI), 8-Chloro-11-piperazin-1-yl-5 H-dibenzo[b,e][1,4 ]diazepine Hydrochloride, Norclozapine HCl |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| LEDER | Cl.Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCNCC4 |
| IUPAC-namn | 3-chloro-6-piperazin-1-yl-11 H-benzo[b][1,4 ]benzodiazepine;hydrochloride |
| Visa mer |