Läs mer
Nordeprenyl (>90%), TRC
Produktkod. 30135226
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135226 25 mg
30003377 5 mg
30007705 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135226 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN66800025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18913-84-3
Molekylformel C12 H15 N
Molekylvikt (g/mol) 173.25
Synonym N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine, rac-Demethylselegiline
IUPAC-namn 1-phenyl-N-prop-2-ynylpropan-2-amine
LEDER CC(Cc1ccccc1)NCC#C
Specifikationer
Kemiskt namn eller material (±)-N-Desmethylselegiline
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C12H15N/c1-3-9-13-11(2)10-12-7-5-4-6-8-12/h1,4-8,11,13H,9-10H2,2H3
Formel vikt 173.1204
Procent renhet >95%