Läs mer
Norethindrone Beta-D-Glucuronide, TRC
Produktkod. 30426640
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30426640 1 mg
1 options
Produktkod. 30426640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN6760051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Norethindrone beta-D-Glucuronide
CAS 64701-11-7
Molekylformel C26H34O8
Kvantitet 1 mg
Synonym (17alpha)-3-Oxo-19-norpregn-4-en-20-yn-17-yl beta-D-Glucopyranosiduronic Acid, Norethindrone Glucuronide
InChI formel InChI=1S/C26H34O8/c1-3-26(34-24-21(30)19(28)20(29)22(33-24)23(31)32)11-9-18-17-6-4-13-12-14(27)5-7-15(13)16(17)8-10-25(18,26)2/h1,12,15-22,24,28-30H,4-11H2,2H3,(H,31,32)/t15-,16+,17+,18-,19-,20-,21+,22-,24-,25-,26-/m0/s1
LEDER C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)C#C
IUPAC-namn (2S,3S,4S,5R,6S)-6-[[(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 474.54
Formel vikt 474.2254