Läs mer
Norethisterone Enanthate, TRC
Produktkod. 30413095
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30413095 100 mg
1 options
Produktkod. 30413095 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN676060100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Norethisterone Enantate (Norethindrone Enanthate)
CAS 3836-23-5
Molekylformel C27 H38 O3
Kvantitet 100 mg
Synonym (17α)-17-[(1-oxoheptyl)oxy]-19-Norpregn-4-en-20-yn-3-one, 19-Nor-17α-pregn-4-en-20-yn-3-one','17-hydroxy-','heptanoate (6CI,7CI,8CI), Heptanoic acid','ester with 17-hydroxy-19-nor-17α-pregn-4-en-20-yn-3-one (8CI), 17α-Ethynyl-19-nortestosterone 17-heptanoate, 17α-Ethynyl-19-nortestosterone enanthate, NSC 22846, Norethindrone enanthate, Norethisterone enanthate, Norethisterone heptanoate, Norigest, Noristerat, SH 8.0393, ZK 5410
InChI formel InChI=1S/C27H38O3/c1-4-6-7-8-9-25(29)30-27(5-2)17-15-24-23-12-10-19-18-20(28)11-13-21(19)22(23)14-16-26(24,27)3/h2,18,21-24H,4,6-17H2,1,3H3/t21-,22+,23+,24-,26-,27-/m0/s1
LEDER CCCCCCC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
IUPAC-namn [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate
Molekylvikt (g/mol) 410.59
Formel vikt 410.2821