Läs mer
Norfloxacin Nicotinate, TRC
Produktkod. 30134390
Change view
Kvantitet:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134390 25 g
30132798 10 g
30006738 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134390 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN68102825G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 118803-81-9
Molekylformel C16H18FN3O3 . C6H5NO2
Molekylvikt (g/mol) 442.4
Synonym Mono[1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylate] 3-Pyridinecarboxylic Acid, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-mono-3-pyridinecarboxylate 3-Quinolinecarboxylic Acid, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid compd. with 3-Pyridinecarboxylic Acid
IUPAC-namn 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;pyridine-3-carboxylic acid
LEDER CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCNCC3.OC(=O)c4cccnc4
Specifikationer
Kemiskt namn eller material Norfloxacin Nicotinate
Kvantitet 25 g
InChI formel InChI=1S/C16H18FN3O3.C6H5NO2/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20;8-6(9)5-2-1-3-7-4-5/h7-9,18H,2-6H2,1H3,(H,22,23);1-4H,(H,8,9)
Formel vikt 442.1652