Läs mer
Norfluoxetine Oxalate, Mikromol™
Produktkod. 30051888
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051888 25 mg
17901086 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051888 Leverantör Mikromol Leverantörsnummer MM0256.180025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Norfluoxetine Oxalate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Norfluoxetine oxalate
CAS 107674-50-0
Analyt- eller komponentnamn Norfluoxetine oxalate
Frakt skick Room Temperature
Molekylformel C16 H16 F3 N O . C2 H2 O4
Certifikat och efterlevnad ISO 17025
Synonym Norfluoxetine Oxalate
InChI formel InChI=1S/C16H16F3NO.C2H2O4/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;3-1(4)2(5)6/h1-9,15H,10-11,20H2;(H,3,4)(H,5,6)
LEDER NCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2.OC(=O)C(=O)O
IUPAC-namn oxalic acid;3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine
Molekylvikt (g/mol) 385.33
Kvantitet 25 mg
Formel vikt 385.1137 g/mol