Läs mer
Norgnoscopine, TRC
Produktkod. 30422359
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30422359 50 mg
1 options
Produktkod. 30422359 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN69990050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Norgnoscopine
CAS 92621-03-9
Molekylformel C21 H21 N O7
Kvantitet 50 mg
Synonym (3S)-6,7-Dimethoxy-3-[(5R)-5,6,7,8-tetrahydro-4-methoxy-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-1(3H)-isobenzofuranone, Norgnoscopine, Nor-α-gnoscopine, N-Desmethyl Noscapine
InChI formel InChI=1S/C21H21NO7/c1-24-12-5-4-11-15(18(12)25-2)21(23)29-17(11)16-14-10(6-7-22-16)8-13-19(20(14)26-3)28-9-27-13/h4-5,8,16-17,22H,6-7,9H2,1-3H3/t16-,17?/m1/s1
LEDER COc1ccc2C(OC(=O)c2c1OC)[C@@H]3NCCc4cc5OCOc5c(OC)c34
IUPAC-namn 6,7-dimethoxy-3-[(5R)-4-methoxy-5,6,7,8-tetrahydro-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-3H-isobenzofuran-1-one
Molekylvikt (g/mol) 399.39
Formel vikt 399.1318