Läs mer
Noribogaine Hydrochloride, TRC
Produktkod. 30423695
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30423695 10 mg
1 options
Produktkod. 30423695 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN69202010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Noribogaine hydrochloride
CAS 110514-35-7
Anteckningar om renhetsgrad HPLC
Molekylformel C19 H24 N2 O . Cl H
Kvantitet 10 mg
Synonym 6,9-Methano-5H-pyrido[1',2':1,2]azepino[4,5-b]indol-2-ol','7-ethyl-6,6a,7,8,9,10,12,13-octahydro-','hydrochloride (1:1)','(6R,6aS,7S,9R,11S)-Ibogaine','O-demethyl-','hydrochloride (6CI), Noribogaine hydrochloride, O-Demethylibogaine hydrochloride
InChI formel InChI=1S/C19H24N2O.ClH/c1-2-12-7-11-8-16-18-14(5-6-21(10-11)19(12)16)15-9-13(22)3-4-17(15)20-18;/h3-4,9,11-12,16,19-20,22H,2,5-8,10H2,1H3;1H/t11-,12+,16+,19+;/m1./s1
LEDER Cl.CC[C@H]1C[C@@H]2C[C@@H]3[C@H]1N(CCc4c3[nH]c5ccc(O)cc45)C2
Molekylvikt (g/mol) 332.87
Formel vikt 332.1655