Läs mer
Norlevo Mepromazine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Desmethyllevomepromazine Hydrochloride |
| CAS | 61733-92-4 |
| Molekylformel | C18 H22 N2 O S . Cl H |
| Kvantitet | 5 mg |
| Synonym | 10H-Phenothiazine-10-propanamine','2-methoxy-N,β-dimethyl-','monohydrochloride','(R)- (9CI), (βR)-2-Methoxy-N,β-dimethyl-10H-phenothiazine-10-propanamine monohydrochloride, (R)-2-Methoxy-N,β-dimethyl-10H-phenothiazine-10-propanamine hydrochloride, Desmethylmethotrimeprazine hydrochloride, N-Demethyllevomepromazine hydrochloride, N-Desmethyllevomepromazine hydrochloride, Norlevomepromazine hydrochloride, Normethotrimeprazine hydrochloride |
| InChI formel | InChI=1S/C18H22N2OS.ClH/c1-13(11-19-2)12-20-15-6-4-5-7-17(15)22-18-9-8-14(21-3)10-16(18)20;/h4-10,13,19H,11-12H2,1-3H3;1H/t13-;/m1./s1 |
| LEDER | Cl.CNC[C@@H](C)CN1c2ccccc2Sc3ccc(OC)cc13 |
| IUPAC-namn | (2R)-3-(2-methoxyphenothiazin-10-yl)-N,2-dimethylpropan-1-amine;hydrochloride |
| Molekylvikt (g/mol) | 350.91 |
| Formel vikt | 350.122 |