Läs mer
Normaprotiline Hydrochloride, TRC
Produktkod. 30410557
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30410557 5 mg
1 options
Produktkod. 30410557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN7170055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Normaprotiline
CAS 5721-37-9
Molekylformel C19 H21 N
Kvantitet 5 mg
Synonym 9,10-Ethanoanthracene-9(10H)-propanamine (9CI), 9,10-Ethanoanthracene-9(10H)-propylamine (8CI), Demethylmaprotiline, Desmethylmaprotiline, N-Desmethylmaprotiline, Normaprotiline, 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine, Maprotiline-desmethyl
InChI formel InChI=1S/C19H21N/c20-13-5-11-19-12-10-14(15-6-1-3-8-17(15)19)16-7-2-4-9-18(16)19/h1-4,6-9,14H,5,10-13,20H2
LEDER NCCCC12CCC(c3ccccc13)c4ccccc24
IUPAC-namn 3-(1-tetracyclo[6.6.2.0^{2,7}.0^{9,14}]hexadeca-2(7),3,5,9(14),10,12-hexaenyl)propan-1-amine
Molekylvikt (g/mol) 263.37674
Formel vikt 263.1674