Läs mer
Northiaden, TRC
Produktkod. 30424791
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424791 5 mg
30411478 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30424791 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN8300505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Northiaden
CAS 1154-09-2
Molekylformel C18 H19 N S
Kvantitet 5 mg
InChI formel InChI=1S/C18H19NS/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18/h2-5,7-11,19H,6,12-13H2,1H3/b16-10-
LEDER CNCC\C=C/1\c2ccccc2CSc3ccccc13
IUPAC-namn (3Z)-3-(6H-benzo[c][1]benzothiepin-11-ylidene)-N-methylpropan-1-amine
Molekylvikt (g/mol) 281.42
Formel vikt 281.1238