Läs mer
Nortropine, TRC
Produktkod. 30417075
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30417075 5 g
1 options
Produktkod. 30417075 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN8457005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 538-09-0
Molekylformel C7 H13 N O
Molekylvikt (g/mol) 127.18
Synonym (3-endo)-8-Azabicyclo[3.2.1]octan-3-ol, 1αH,5αH-Nortropan-3α-ol (8CI), 8-Azabicyclo[3.2.1]octan-3-ol, endo-, Nortropine (6CI,7CI), (3-endo)-8-Azabicyclo[3.2.1]octan-3-ol, 3α-Nortropanol, N-Demethyltropine, NSC 72850, Nortropenol, Nortropin, Tropigenin, Tropigenine, endo-8-Azabicyclo[3.2.1]octan-3-ol
IUPAC-namn (1S,5R)-8-azabicyclo[3.2.1]octan-3-ol
LEDER O[C@H]1C[C@H]2CC[C@@H](C1)N2
Specifikationer
Kemiskt namn eller material Nortropine
Kvantitet 5 g
InChI formel InChI=1S/C7H13NO/c9-7-3-5-1-2-6(4-7)8-5/h5-9H,1-4H2/t5-,6+,7+
Formel vikt 127.0997