Läs mer
Norvancomycin hydrochloride, MedChemExpress
Produktkod. 16488826
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16488826 100 mg 100mg
16484637 50 mg -
16468728 10 mg -
16461138 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16488826 Leverantör Medchem Express Leverantörsnummer HYB1924
MedChemExpress Norvancomycin hydrochloride is applicable for endocarditis, osteomyelitis, pneumonia, sepsis or soft tissue infections caused by Staphylococcus (including Methicillin-resistant strains and multidrug-resistant microbial strains).
Norvancomycin hydrochloride is applicable for endocarditis, osteomyelitis, pneumonia, sepsis or soft tissue infections caused
Specifikationer
Kemiskt namn eller material Norvancomycin hydrochloride
CAS 213997-73-0
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C65H74Cl3N9O24
Kvantitet 100 mg
Synonym Desmethyl-vancomycin hydrochloride
Löslighetsinformation H2O : ≥ 100 mg/mL (67.95 mM) ∣DMSO : ≥ 100 mg/mL (67.95 mM)
LEDER O[C@@H]1[C@@H](O[C@@](O[C@@H](C)[C@H]2O)([H])C[C@@]2(N)C)[C@@H](O[C@H](CO)[C@H]1O)OC3=C(OC4=CC=C([C@H]([C@H]5NC([C@H](N)CC(C)C)=O)O)C=C4Cl)C=C([C@](NC([C@@H](NC5=O)CC(N)=O)=O)([H])C(N[C@](C6=O)([H])C7=CC8=C(O)C=C7)=O)C=C3OC9=CC=C([C@H]([C@](C(N[C@@](C(O)=O)([H])C%10=C8C(O)=CC(O)=C%10)=O)([H])N6)O)C=C9Cl.Cl[H]
Molekylvikt (g/mol) 1471.69
Formel vikt 1471.69
Procent renhet 96.79%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation