Learn More
NOS1AP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563147
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KKKSQAMRIV RTVGQAFEVC HKLSLQHTQQ NADGQEDGES ERNSNSSGDP GRQLTGAERA STATAEETDI DAVEVPLPGN DV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
NOS1AP | |
Polyclonal | |
Unconjugated | |
NOS1AP | |
6330408P19Rik; Capon; carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein; C-terminal PDZ domain ligand of neuronal nitric oxide synthase; C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON); C-terminal PDZ ligand of neuronal nitric oxide synthase protein; Kiaa0464; ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain; MGC138500; nitric oxide synthase 1 (neuronal) adaptor protein; nitric oxide synthase 1 adaptor protein; NOS1AP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9722 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75052 | |
NOS1AP | |
Recombinant protein corresponding to Human NOS1AP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only