Learn More
iNOS Monoclonal Antibody (CXNFT), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1376.00 SEK - 3345.00 SEK
Specifications
Antigen | iNOS |
---|---|
Clone | CXNFT |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15530227
|
Affymetrix eBioscience
53-5920-80 |
25 μg |
1376.00 SEK
25µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15540227
|
Affymetrix eBioscience
53-5920-82 |
100 μg |
3345.00 SEK
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This CXNFT monoclonal antibody reacts to mouse NOS2 (inducible NOS, iNOS). Nitric oxide synthase enzymes catalyze the formation of nitric oxide from L-arginine through an NADPH- and oxygen-dependent mechanism. There are three isoforms of NOS that are encoded by three separate genes. NOS1 (neuronal NOS, nNOS) and NOS3 (endothelial NOS, eNOS) are constitutively expressed, while NOS2 is induced in response to bacterial endotoxins and inflammatory cytokines such as IFN gamma and TNF alpha. NOS2 is expressed by myeloid-derived suppressor cells and M1 macrophages but not alternatively activated M2 macrophages. NOS enzymes are functionally active only when they form homodimers, and dimerization of NOS2 occurs at steady-state concentrations of free Ca2+ such that NOS2 is functionally active when it is produced. Applications Reported: This CXNFT antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This CXNFT antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse thioglycolate-elicited peritoneal exudate cells using the intracellular Fixation and Permeabilization Buffer Set (cat. 88-8824) and protocol. The Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) may also be used with similar results. This can be used at less than or equal to 0.06 μg per test.
iNOS Monoclonal antibody specifically detects iNOS in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
iNOS | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2 | |
Nos2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Nos2 |
CXNFT | |
Flow Cytometry, Immunocytochemistry, Western Blot | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
P29477 | |
18126 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.