Läs mer
Noscapine N-Oxide Hydrochloride, TRC
Produktkod. 30425037
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30425037 50 mg
1 options
Produktkod. 30425037 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN88202050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Noscapine N-Oxide Hydrochloride
CAS 201015-28-3
Molekylformel C22 H23 N O8 . H Cl
Kvantitet 50 mg
Synonym [S-(R*,S*)]-6,7-Dimethoxy-3-(5,6,7,8-tetrahydro-4-methoxy-6-methyl-6-oxido-1,3-dioxolo[4,5-g]isoquinolin-5-yl)-1(3H)-isobenzofuranone Hydrochloride, Narcotine N-oxide Hydrochloride, α-Narcotine N-Oxide Hydrochloride
InChI formel InChI=1S/C22H23NO8.ClH/c1-23(25)8-7-11-9-14-20(30-10-29-14)21(28-4)15(11)17(23)18-12-5-6-13(26-2)19(27-3)16(12)22(24)31-18;/h5-6,9,17-18H,7-8,10H2,1-4H3;1H/t17-,18+,23?;/m1./s1
LEDER Cl.COc1ccc2[C@H](OC(=O)c2c1OC)[C@H]3c4c(CCN3(=O)C)cc5OCOc5c4OC
IUPAC-namn (3S)-6,7-dimethoxy-3-[(5R)-4-methoxy-6-methyl-6-oxo-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-3H-isobenzofuran-1-one;hydrochloride
Molekylvikt (g/mol) 465.88
Formel vikt 465.119