Läs mer
Nosiheptide (>85%), TRC
Produktkod. 30134793
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134793 50 mg
17941385 1 mg
17974175 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134793 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN88400050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56377-79-8
Molekylformel C51 H43 N13 O12 S6
Molekylvikt (g/mol) 1222.36
Synonym Nosiheptide, 30,32-Imino-8,5:18,15:40,37-trinitrilo-21,36-([2,4]-endo-thiazolomethanimino)-5H,15H,37H-pyrido[3,2-w][2,11,21,27,31,7,14,17]benzoxatetrathiatriazacyclohexatriacontine, nosiheptide deriv., N-[1-(Aminocarbonyl)ethenyl]-2-[(11S,14Z,21S,23S,29S)-14-ethylidene-9,10,11,12,13,14,19,20,21,22,23,24,26,33,35,36-hexadecahydro-3,23-dihydroxy-11-[(1R)-1-hydroxyethyl]-31-methyl-9,12,19,24,33,43-hexaoxo-30,32-imino-8,5:18,15:40,37-trinitrilo-21,36-([2,4]-endo-thiazolomethanimino)-5H,15H,37H-pyrido[3,2-w][2,11,21,27,31,7,14,17]benzoxatetrathiatriazacyclohexatriacontin-2-yl]-4-thiazolecarboxamide, Multhiomycin, Multiomycin, NSC 307240, Primofax, RP 9671, [11S-[11R*(S*),14Z,21R*,23R*,29R*]]-N-[1-(aminocarbonyl)ethenyl]-2-[14-ethylidene-9,10,11,12,13,14,19,20,21,22,23,24,26,33,35,36-hexadecahydro-3,23-dihydroxy-11-(1-hydroxyethyl)-31-methyl-9,12,19,24,33,43-hexaoxo-30,32-imino-8,5:18,15:40,37-trinitrilo-21,36-([2,4]-endo-thiazolomethanimino)-5H,15H,37H-pyrido[3,2-w][2,11,21,27,31,7,14,17]benzoxatetrathiatriazacyclohexatriacontin-2-yl]-4-thiazolecarboxamide
LEDER C\C=C\1/NC(=O)C(NC(=O)c2csc(n2)c3cc(O)c(nc3c4csc(n4)C5CSC(=O)c6[nH]c7cccc(COC(=O)C(O)CC(NC(=O)c8csc1n8)c9nc(cs9)C(=O)N5)c7c6C)c%10nc(cs%10)C(=O)NC(=C)C(=O)N)C(C)O
Specifikationer
Kemiskt namn eller material Nosiheptide
Kvantitet 50 mg
InChI formel InChI=1S/C51H43N13O12S6/c1-5-23-46-60-28(14-79-46)41(70)56-25-10-33(67)50(74)76-11-21-7-6-8-24-34(21)18(2)35(54-24)51(75)82-17-31(57-42(71)29-15-80-47(25)61-29)48-58-26(12-78-48)37-22(45-59-30(13-77-45)43(72)64-36(20(4)65)44(73)55-23)9-32(66)38(63-37)49-62-27(16-81-49)40(69)53-19(3)39(52)68/h5-9,12-16,20,25,31,33,36,54,65-67H,3,10-11,17H2,1-2,4H3,(H2,52,68)(H,53,69)(H,55,73)(H,56,70)(H,57,71)(H,64,72)/b23-5-
Formel vikt 1221.1478