Learn More
NOSTRIN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560695
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TLENCYQSIL ELEKERIQLL CNNLNQYSQH ISLFGQTLTT CHTQIHCAIS KIDIEKDIQA VMEETAILST ENKSEFLLTD YFEEDPNSAM DKERRKSLLK PKLLRL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.
NOSTRIN Polyclonal antibody specifically detects NOSTRIN in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NOSTRIN | |
Polyclonal | |
Unconjugated | |
Nostrin | |
BM247; BM247 homolog; Dab2-interacting protein 2; Daip2; Disabled homolog 2-interacting protein 2; endothelial nitric oxide synthase traffic inducer; ENOS traffic inducer; eNOS trafficking inducer; eNOS-trafficking inducer; mDaIP2; Nitric oxide synthase traffic inducer; nitric oxide synthase trafficker; nitric oxide synthase trafficking; nostrin; ortholog of mouse disabled 2 interacting proein 2; probable ortholog of human nitric oxide synthase trafficker (NOSTRIN) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVI9 | |
Nostrin | |
Recombinant protein corresponding to Human NOSTRIN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only