missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NOTCH2NL Recombinant Protein
Recombinant protein for NOTCH2NL (human) gene
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH19835 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 388677 |
Molecular Weight (g/mol) | 51.7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180494
|
Abnova™
H00388677-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190494
|
Abnova™
H00388677-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Human NOTCH2NL full-length ORF ( AAH19835, 1 a.a. - 236 a.a.) recombinant protein with GST-tag at N-terminal
- Theoretical molecular weight: 51.7kDa
- Preparation: in vitrowheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer
Sequence: MCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVGFTGKECQWTDACLSHPCANGSTCTTVANQFSCKCLTGFTGQKCETDVNECDIPGHCQHGGICLNLPGSYQCQCLQGFTGQY CDSLYVPCAPSPCVNGGTCRQTGDFTFECNCLPETVRRGTELWERDREVWNGKEHDEN
Best use within three months from the date of receipt.
ELISA, western blotting (recombinant protein), antibody production, protein array
Specifications
AAH19835 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
51.7 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NOTCH2NL | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
388677 | |
NOTCH2NL (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVGFTGKECQWTDACLSHPCANGSTCTTVANQFSCKCLTGFTGQKCETDVNECDIPGHCQHGGICLNLPGSYQCQCLQGFTGQYCDSLYVPCAPSPCVNGGTCRQTGDFTFECNCLPETVRRGTELWERDREVWNGKEHDEN | |
N2N | |
NOTCH2NL | |
Wheat Germ (in vitro) | |
GST |