Learn More
Abnova™ NOTCH2NL Recombinant Protein
Recombinant protein for NOTCH2NL (human) gene
Brand: Abnova™ H00388677-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Human NOTCH2NL full-length ORF ( AAH19835, 1 a.a. - 236 a.a.) recombinant protein with GST-tag at N-terminal
- Theoretical molecular weight: 51.7kDa
- Preparation: in vitrowheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer
Sequence: MCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVGFTGKECQWTDACLSHPCANGSTCTTVANQFSCKCLTGFTGQKCETDVNECDIPGHCQHGGICLNLPGSYQCQCLQGFTGQY CDSLYVPCAPSPCVNGGTCRQTGDFTFECNCLPETVRRGTELWERDREVWNGKEHDEN
Best use within three months from the date of receipt.
ELISA, western blotting (recombinant protein), antibody production, protein array
Specifications
AAH19835 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
51.7 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVGFTGKECQWTDACLSHPCANGSTCTTVANQFSCKCLTGFTGQKCETDVNECDIPGHCQHGGICLNLPGSYQCQCLQGFTGQYCDSLYVPCAPSPCVNGGTCRQTGDFTFECNCLPETVRRGTELWERDREVWNGKEHDEN | |
N2N | |
NOTCH2NL | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
388677 | |
NOTCH2NL (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NOTCH2NL | |
Human | |
Recombinant | |
Solution |