missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ NOTCH4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540587
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: LLLGLGAARE LRDQAGLAPA DVAHQRNHWD LLTLLEGAGP PEARHKATPG Sequence homology: Cow: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Pig: 100%; Rat: 93%.
This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. This gene may be associated with susceptibility to schizophrenia in a small portion of cases (entrez gene).
Specifikationer
| NOTCH4 | |
| Polyclonal | |
| Unconjugated | |
| NOTCH4 | |
| DAQB-143M3.4; FLJ16302; hNotch4; Int3; Int-3; MGC74442; N4; neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch gene homolog 4; Notch homolog 4; notch receptor 4; NOTCH3; Notch4; putative Notch4 protein; Transforming protein Int-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4855 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99466 | |
| NOTCH4 | |
| Synthetic peptide directed towards the middle region of human NOTCH4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering