Learn More
Merck Millipore Novagen™ KOD Hot Start DNA Polymerase
For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications
Brand:  Merck Millipore 71086-3
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12838435
  • 6585.00 SEK / Each
Estimated Shipment: 27-06-2024
to see stock.
Description
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
Special Offers
For Research Use Only