Learn More
Merck Millipore Novagen™ KOD Hot Start Master Mix
For convenient high fidelity PCR
POA
Description
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA template up to 21 kbp
  • Eliminates mispriming and primer-dimer formation
  • Convenient ambient-temperature setup compatible with automation
  • KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets