Learn More
EMD Millipore™ Novagen™ KOD Hot Start Master Mix
For convenient high fidelity PCR
Brand:  EMD Millipore™ 71842-3
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15448911
  • 2694.00 SEK / 100 reactions
Estimated Shipment
25-09-2018
Add to basket
Description
Description
  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA template up to 21 kbp
  • Eliminates mispriming and primer-dimer formation
  • Convenient ambient-temperature setup compatible with automation
  • KOD Hot Start Buffer ensures optimal PCR performance over wide range of targets
Specifications
Specifications
KOD Hot Start Master Mix
100 rxn
-20°C