Learn More
Merck Millipore Novagen™ Overnight Express™ Autoinduction System 1
Induces protein expression without monitoring cell density.
Brand:  Merck Millipore 71300-4
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15216474
  • 4670.00 SEK / 5 L
Estimated Shipment
15-10-2020
Add to basket
Description
Description
  • Blend of carbon sources optimized for tightly regulated uninduced growth to high cell density followed by high-level induction
  • Provides magnesium for maximal cell density
  • Convenient for routine expression of proteins in multiple cultures
  • Ideal for high-throughput parallel analysis of protein expression, solubility, purification from multiple expression clones
  • Allows rapid optimization of the host-vector combinations, expression conditions
  • Purification parameters for high-throughput production of proteins for structural or functional analysis
  • Packaging: Glass Bottle
Specifications
Specifications
5L
15°C to 30°C