Learn More
Invitrogen™ Novex™ 12% Tris-Glycine Protein Gel, 26-well
Separation Range: 20 to 250 kd; Separation Type: Denaturing, Native; For Use With: Midi Gel Tank, XCell4 SureLock Midi
Brand: Invitrogen™ WT0123BOX
Additional Details : Weight : 0.98000kg
Includes: One box of 10 gels
Description
- Provide reproducible separation of a wide range of proteins into well-resolved bands
- Individually packaged gels for optimal experiment design
- Compatible with most protein standards
- Usable for native and denaturing protein assays
- Do not contain SDS and can be used to run proteins in native or in denatured form
Sample Loading Volume: 15μL; 1D Well
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
XCell4 SureLock Midi-Cell | |
Tris-Glycine | |
Protein | |
Denaturing, Native | |
1 Box | |
8 cm |
12% | |
Homogeneous Page | |
20 to 250 kDa | |
26-well | |
1D Well | |
13 cm |
For Research Use Only. Not for use in diagnostic procedures.