Learn More
Invitrogen™ Empty Gel Cassette Combs, mini, 1.5 mm, 15 well
Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)
Brand:  Invitrogen™ NC3515
Additional Details : Weight : 0.00454kg
Product Code. 10185542
  • 455.00 SEK / Pack of 25
Estimated Shipment: 29-04-2024
to see stock.
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
These combs are for use when pouring special gel types and are available in two thicknesses (1.0mm and 1.5mm) and seven well formats (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis