missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Empty Gel Cassette Combs, Mini

Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)

362.00 SEK

Specifications

For Use With (Equipment) Mini-size Empty Gel Cassettes
View More Specs

Products 2
Product Code Brand Type Thickness (Metric) Price Quantity & Availability  
Product Code Brand Type Thickness (Metric) Price Quantity & Availability  
10137552
View Documents
Invitrogen™
NC3002
2D-well Empty Gel Cassette Comb 1.0 mm 362.00 SEK
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment
17-11-2020
Add to basket
 
10338802
View Documents
Invitrogen™
NC3003
2-well Empty Gel Cassette Comb 1.0 mm 362.00 SEK
Pack of 25 View alternative products
Save Up to 
»
Estimated Shipment
06-10-2020
Add to basket
 
Description

Description

These combs are for use when pouring special gel types and are available in two thicknesses (1.0mm and 1.5mm) and seven well formats (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).

Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Specifications

Mini-size Empty Gel Cassettes

For Research Use Only. Not for use in diagnostic procedures.