missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Gel Loading Tips

For easy and accurate loading of samples

592.00 SEK - 1266.00 SEK

Specifications

Type Gel Loading Tips
View More Specs

Includes: One pack (200/pack) of Standard, Round gel loading tips.

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11594986 View Documents Invitrogen™
LC1002
Flat Tips 1266.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 2
Add to basket
 
10422522 View Documents Invitrogen™
LC1001
Standard round Tips 592.00 SEK
Pack of 200 View alternative products
Save Up to 
»
EU Stock 120
Add to basket
 
Description

Description

Tips are RNase/DNase-free and non-autoclavable. Standard, round can be used for easy and accurate loading of samples onto 1.0mm and 1.mm gels. Loading volume is 0.5 to 200 μL.

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Specifications

Gel Loading Tips

For Research Use Only. Not for use in diagnostic procedures.