Läs mer
Invitrogen™ Large Gel Drying Kit
Produktkod. 10573103
Change view
Kvantitet:
1 sats
Förpackningsstorlek:
5ml
Produktkod. Kvantitet Förpackningsstorlek
10573103 1 sats 5ml
1 options
Produktkod. 10573103 Leverantör Invitrogen™ Leverantörsnummer NI2207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Innehåller
DryEase™ Large Gel Drying Frame (1 frame, 8 clamps), DryEase™ Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500mL).
Accommodate larger format gels or four mini-gels
The Large Gel Drying Kit dries gels using the same components as the DryEase™ Mini-Gel Drying System. Consistently dry gels without cracking and can accommodate up to four mini-gels or a large format gel. Base not required for assembly.
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring The Large Gel Drying Kit contains a DryEase™ Large Gel Drying Frame (1 frame, 8 clamps), DryEase™ Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500 ml). Store solution at room temperature. Guaranteed stable for one year when properly stored.
Kvantitet 1 sats
Produktlinje Novex
Produkttyp Large Gel Drying Kit