missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ XCell SureLock™ Replacement Parts

For the XCell SureLock Mini Blot Module

260.00 SEK - 2640.00 SEK

Specifications

For Use With (Equipment) XCell SureLock Mini Blot Module
View More Specs

Products 8
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10369272 View Documents Invitrogen™
EI0012
Buffer Dam 359.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 9
Add to basket
 
10553483 View Documents Invitrogen™
EI0010
Lid 1556.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 5
Add to basket
 
10277112 View Documents Invitrogen™
EI0011
Wedge 1042.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 3
Add to basket
 
10184962 View Documents Invitrogen™
EI0013
Lower Buffer Chamber 2640.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 7
Add to basket
 
10770584 View Documents Invitrogen™
EI9014
Buffer Core 2390.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 1
Add to basket
 
10729433 View Documents Invitrogen™
EI9022
Platinum Wire 328.00 SEK
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-02-2020
Add to basket
 
10257402 View Documents Invitrogen™
EI9010
Cassette Opener 260.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 36
Add to basket
 
11554756 View Documents Invitrogen™
EI9052
Sponge Pad 394.00 SEK
Pack of 8 View alternative products
Save Up to 
»
EU Stock 37
Add to basket
 
Description

Description

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Room Temperature

Specifications

Specifications

XCell SureLock Mini Blot Module

For Research Use Only. Not for use in diagnostic procedures.