Läs mer
Novus Biologicals™ Recombinant Human HLA C His Protein
Produktkod. 18796913 Handla allt Bio Techne Produkter
Kvantitet:
0.1 mg
Förpackningsstorlek:
0.1mg
Produktkod. 18796913
Brand: Novus Biologicals™ NBP223210
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-308 of Human HLA C The Recombinant Human HLA C Protein is derived from E. coli. The Recombinant Human HLA C Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 0.1 mg