Läs mer
Novus Biologicals™ pCL-Ampho Retrovirus Packaging Vector
Applications: Retroviral Production
Brand: Novus Biologicals™ NBP2-29541
Beskrivning
The pCL-Ampho packaging vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. By introducing a retroviral vector into a cell expressing retroviral proteins, retroviral particles (virions) are shed into the culture medium at the rate of 1 infectious particle/cell/day. Retrovirus tropism is determined at three levels. The first is simply a function of viral envelope protein, gp70. The envelope determines which cells the virus will enter. gp70 comes in three different flavors for gene therapy. The plasmid DNA can be amplified by transforming bacteria and growing them on LB/ampicillin (100 ug/ml) media.
- Species: Human
- Specificity: Ectotropic (usually) (MoMuLV) Mouse and rat cells only (not human), Amphotropic (from 4070A MuLV) Most mammalian cells (but not hamster) 10A1 (MuLV) Most mammalian cells (including hamster)
- Buffer: 10µg in 20µL sterile water
Rekommenderad förvaring
Store at -20°C. Avoid freeze-thaw cycles.
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.