Läs mer
NOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifikationer
| Antigen | NOX1 |
|---|---|
| Koncentration | 0.23 mg/mL |
| Användningsområden | Western Blot, ELISA |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
16877744
|
Proteintech
17772-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
16867744
|
Proteintech
17772-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.Specifikationer
| NOX1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8CIZ9, Q9WV87, Q9Y5S8 | |
| 114243, 237038, 27035 | |
| NOX1 Fusion Protein Ag12187 | |
| Primary | |
| -20°C | |
| NOX1 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GP91 2, Mitogenic oxidase 1, MOX 1, MOX1, NADPH oxidase 1, NOH 1, NOH1, NOX 1, NOX1 | |
| Nox1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |