missing translation for 'onlineSavingsMsg'
Learn More

NOX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20065-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16879544

  • 1236.20 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to NOX3.

The NOX family of NAPDH oxidases is comprised of seven transmembrane proteins that oxidize intracellular NAPDH/NADH, causing electron transport across the membrane and the reduction of molecular oxygen to superoxide. NOX3 is expressed predomitly in the inner ear and is involved in the biogenesis of otoconia/otolith. It has been suggested that NOX3 is activated by the Transient Receptor Potential Vanilloid 1 (TRPV1), and this activity causes increased levels of reactive oxygen species in the inner ear, which in turn leads to STAT1-mediated inflammation and hearing loss.
Specifications

Specifications

NOX3
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q672J9, Q9HBY0
GP91 3, gp91phox homolog 3, Mitogenic oxidase 2, MOX 2, MOX2, NADPH oxidase 3, NOX3
Rabbit
IgG
20 μL
-20°C
Primary
224480, 50508
Western Blot
Unconjugated
NOX3
Liquid
NOX3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers