missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | NOX3 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879544
|
Proteintech
20065-1-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16869544
|
Proteintech
20065-1-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The antibody is specific to NOX3.
The NOX family of NAPDH oxidases is comprised of seven transmembrane proteins that oxidize intracellular NAPDH/NADH, causing electron transport across the membrane and the reduction of molecular oxygen to superoxide. NOX3 is expressed predomitly in the inner ear and is involved in the biogenesis of otoconia/otolith. It has been suggested that NOX3 is activated by the Transient Receptor Potential Vanilloid 1 (TRPV1), and this activity causes increased levels of reactive oxygen species in the inner ear, which in turn leads to STAT1-mediated inflammation and hearing loss.Specifications
NOX3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q672J9, Q9HBY0 | |
224480, 50508 | |
Peptide | |
Primary | |
-20°C | |
NOX3 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GP91 3, gp91phox homolog 3, Mitogenic oxidase 2, MOX 2, MOX2, NADPH oxidase 3, NOX3 | |
NOX3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |