Läs mer
NPEC-caged-noradrenalin, TRC
Produktkod. 30414093
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30414093 10 mg -
30411565 1 mg -
30411999 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30414093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN89902010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1257323-83-3
Molekylformel C17H18N2O7
Molekylvikt (g/mol) 362.33
LEDER O=C(OC(C1=CC=CC=C1[N+]([O-])=O)C)NC[C@@H](C2=CC=C(O)C(O)=C2)O
Specifikationer
Kemiskt namn eller material NPEC-caged-noradrenalin
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C17H18N2O7/c1-10(12-4-2-3-5-13(12)19(24)25)26-17(23)18-9-16(22)11-6-7-14(20)15(21)8-11/h2-8,10,16,20-22H,9H2,1H3,(H,18,23)/t10?,16-/m0/s1
Formel vikt 362.1114
Procent renhet >95%