missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NPM1 (Human) Recombinant Protein
Human NPM1 full-length ORF ( AAH02398.1, 1 a.a. - 294 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004869-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEDSMDMDMSPLRPQNYLFGCELKADKDYHFKVDNDENEHQLSLRTVSLGAGAKDELHIVEAEAMNYEGSPIKVTLATLKMSVQPTVSLGGFEITPPVVLRLKCGSGPVHISGQHLVAVEEDAESEDEEEEDVKLLSISGKRSAPGGGSKVPQKKVKLAADEDDDDDDEEDDDEDDDDDDFDDEEAEEKAPVKKSIRDTPAKNAQKSNQNGKDSKPSSTPRSKGQESFKKQEKTPKTPKGPSSVEDIKAKMQASIEKGGSLPKVEAKFINYVKNCFRMTDQEAIQDLWQWRKSL
Specifications
AAH02398.1 | |
nucleophosmin (nucleolar phosphoprotein B23, numatrin) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NPM1 | |
GST |
4869 | |
Wheat germ expression system | |
25 μg | |
B23, MGC104254, NPM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |