missing translation for 'onlineSavingsMsg'
Learn More

NPPA Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27426-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16875615

  • 4721.67 SEK / 150µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.
Specifications

Specifications

NPPA
Polyclonal
Unconjugated
NPPA
ANF, ANP, ATFB6, Atrial natriuretic factor, Atrial natriuretic peptide, CDD ANF, CDP, NPPA, PND, Prepronatriodilatin
Rabbit
Antigen Affinity Chromatography
RUO
230899, 4878
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01160, P05125
NPPA
NPPA Fusion Protein Ag26692
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers