missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26706-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
NPR3 | |
Polyclonal | |
Unconjugated | |
NPR3 | |
ANP C, ANPR C, ANPRC, GUCY2B, NPR3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4883 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17342 | |
NPR3 | |
NPR3 Fusion Protein Ag19071 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |