missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRCAM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21608-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Cell adhesion, ankyrin-binding protein involved in neuron-neuron adhesion. May play a role in the molecular assembly of the nodes of Ranvier.Specifications
NRCAM | |
Polyclonal | |
Unconjugated | |
NRCAM | |
hBravo, KIAA0343, neuronal cell adhesion molecule, Neuronal surface protein Bravo, Ng CAM related, Nr CAM, NRCAM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
319504, 4897, 497815 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97686, Q810U4, Q92823 | |
Nrcam | |
NRCAM Fusion Protein Ag16237 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |